Learn More
Agroclavine, TRC
Product Code. 30134572
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134572 10 mg 10mg
30135307 25 mg 25mg
17990535 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134572 Supplier Toronto Research Chemicals Supplier No. TRCA43004010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 548-42-5
Molecular Formula C16 H18 N2
Molecular Weight (g/mol) 238.33
Synonym Ergoline, 8,9-didehydro-6,8-dimethyl-, Agroclavine (6CI), 8,9-Didehydro-6,8-dimethylergoline, Indolo[4,3-fg]quinoline, ergoline deriv., (-)-Agroclavine, Agroclavin, NSC 93132
IUPAC Name (6aR,10aR)-7,9-dimethyl-6,6a,8,10a-tetrahydro-4H-indolo[4,3-fg]quinoline
SMILES CN1CC(=C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34)C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C16H18N2/c1-10-6-13-12-4-3-5-14-16(12)11(8-17-14)7-15(13)18(2)9-10/h3-6,8,13,15,17H,7,9H2,1-2H3/t13-,15-/m1/s1
Formula Weight 238.147
Percent Purity >95%
Chemical Name or Material Agroclavine
Grade HPLC