missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF Antibody (4B2), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | AIF |
|---|---|
| Clone | 4B2 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043187
|
Novus Biologicals
NBP3-26515-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043871
|
Novus Biologicals
NBP3-26515-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
AIF Monoclonal antibody specifically detects AIF in Human, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistryEspecificaciones
| AIF | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
| A synthesized peptide derived from Human AIF [UniProt O95831] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 4B2 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Inhibitors Activators and Regulators, Mitochondrial Markers, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 9131 | |
| IgG | |
| Affinity purified |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto