Learn More
BINDER™ Measurement of Air Exchange Rate Instrument
BINDER™ Measurement of Air Exchange Rate Instrument is designed according to ASTM D5374.
Brand:  BINDER™ 8012-0195
Additional Details : Weight : 0.01000kg
Product Code. 15631686
  • 1200.00€ / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Accessory for Drying and heating chamber
  • Measures exchange rate in ambient temperature
  • Measurement in factory
  • With definition and protocol according to ambient temperature
  • For use with all FB and M models
Specifications
Specifications
Air Exchange Measurement