Learn More
Abeba™ Air-Cushion Sole 2610 Shoes
Antistatic with air cushion PU-sole and high slip resistance. Sanitized™ treated.
Brand:  Abeba™ 2610/44
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15628157
  • 87.63€ / 1 pair
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O2 SRA
  • Smooth leather
  • Black
  • Slip resistance PU outsole sole
  • Replaceable insole (article no. 3551)
  • Sanitized treated
  • Antistatic
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20347:2012 O2 SRA
Microfiber
44
Polyurethane
Black
Leather
Slip-resistant
2610