Learn More
Aalborg™ Air Direct Glass Flow Tube 65mm Reference Scale
Product Code. 11892450
Change view
Click to view available options
Airflow Rate:
10 L/min.
100 mL/min.
1000 mL/min.
104 mL/min.
1946 mL/min.
2 L/min.
2.5 L/min.
2000 mL/min.
24.6 L/min.
250 mL/min.
299 mL/min.
4 L/min.
40 L/min.
4922 mL/min.
5 L/min.
50 mL/min.
500 mL/min.
5000 mL/min.
6 mL/min.
62 L/min.
75 mL/min.
986 mL/min.
Unit Size:
Each
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
11892450 40 L/min. Each
11812430 50 mL/min. Each
11852450 2000 mL/min. Each
11872450 10 L/min. Each
11802320 4922 mL/min. Each
11872310 2 L/min. Each
11882310 4 L/min. Each
11862450 5 L/min. Each
11802300 104 mL/min. Each
11872430 100 mL/min. Each
11862310 2.5 L/min. Each
11832440 250 mL/min. Each
11862290 6 mL/min. Each
11832310 986 mL/min. Each
11842310 1946 mL/min. Each
11822450 1000 mL/min. Each
11852330 24.6 L/min. Each
11842430 75 mL/min. Each
11882330 62 L/min. Each
11812300 299 mL/min. Each
11768328 5000 mL/min. Each
11812450 500 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Product Code. 11892450 Supplier Aalborg™ Supplier No. FT06403STVN
  • ONLINE EXCLUSIVE PRICE
    93.45€
    57.12€ / Pack of 1
    Save 36.33€
    38% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Aalborg™ Air Direct Glass Flow Tube Reference Scale reults in superior mL and L readings by providing a scale of meaurements in a vairety of volumes.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Type Air Direct Flow Tube
Width (Metric) 65 mm
Airflow Rate 40 L/min.