Learn More
Aalborg™ Air Mass Flowmeter
Product Code. 11713479
Change view
Click to view available options
Airflow Rate:
0 to 1 LPM
0 to 10 LPM
0 to 100 mL/min.
0 to 2 LPM
0 to 20 mL/min.
0 to 5 LPM
0 to 50 mL/min.
0 to 500 mL/min.
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate unitSize
11713479 0 to 20 mL/min. Each
11717839 0 to 100 mL/min. Each
11747829 0 to 50 mL/min. Each
11822280 0 to 500 mL/min. Each
11872170 0 to 1 LPM Each
11832220 0 to 2 LPM Each
11892170 0 to 10 LPM Each
11882170 0 to 5 LPM Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11713479 Supplier Aalborg™ Supplier No. GFC17AVAL6C0/3AB02SS
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Type Mass Flowmeter
Accuracy ±5%
Airflow Rate 0 to 20 mL/min.
Certifications/Compliance NIST calibration certificate
Power Supply Details 12 V to 200 mA max
Precision ±1.5 %
Scale Full scale
Temperature (Metric) 5°C to 50°C