Learn More
Asecos™ Air Purifier PURIFIAIR.620
Product Code. 17222534 Shop All asecos Products
Change view
Click to view available options
Product Type:
Replacement Filter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
17222534 Replacement Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17222534 Supplier Asecos™ Supplier No. 39326
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
Air Purifier PURIFIAIR.620 reduces indoor viral loads and pollutant levels with a multi-component filter system.
  • Asecos PURIFIAIR.620 can significantly reduce viral loads and pollution in indoor areas
    • Removes mould spores, pollen, and allergens
    • Eliminates germs and bacteria and neutralises them
    • Continuously filters out fine dust particles
    • Removes unpleasant odours and harmful gases from indoor atmospheres
    • Built-in PM1.0 sensor and colour display allow users to quickly identify and read off the indoor atmosphere particle concentration in four levels from green (low) to red (high) concentration
    • Built-in combination of pre-filters, an E12 EPA Filter and H14 HEPA Filter in accordance with EN 1822 has an efficiency of up to 99.995% while keeping out particles from 0.1 bis 0.3 μm in size
    • H14 HEPA filters are used in areas where protection against infection plays a key role
    • The mobile design allows the purifier to be used flexibly in different areas
  • 1 device handles a volume flow rate of 160 bis to 620 m³/h (5 operating levels) and is suitable for approx. 100 m³ room volume (corresponding to a surface area of 40 m² and a room height of 2.5 m)
    • A larger number of devices should be used in larger rooms
    • Fresh air must be supplied to the rooms via natural or technical ventilation
    • Air purifiers should not be considered a substitute for a fresh air supply
    • Efficient results over time only if the filter is replaced at regularly according to the electronic display
    • Large filter units ensure a long service life
    • Low energy requirements result in low usage costs
Specifications
Filter Type Prefilter, Cold Catalysis Filter, Activated Carbon Filter, High-performance Particle Filter
For Use With (Equipment) AIRPURIFIAIR.620
Weight (Metric) 2.867 Kg
Height (Metric) 162 mm
Quantity 1
Depth (Metric) 509 mm
Width (Metric) 402 mm
Product Type Replacement Filter
Certifications/Compliance EN 1822