Learn More
Syngene™ AirClean™ Microscope Enclosure with UVTect™ Microprocessor Controller
Product Code. 17735904 Shop All Syngene Products
Change view
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Unit Size
17735904 32 in. Each
17705914 48 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17735904 Supplier Syngene™ Supplier No. SYNAC632TLFUVCMIC220E
  • ONLINE EXCLUSIVE PRICE
    11780.00€
    9897.78€ / Pack of 1
    Save 1882.22€
    15% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The downflow AirClean™ Microscope Enclosure provides a clean work environment and a UV light for irradiation between processes. UVTect™ microprocessor controller provides one-touch operation of all enclosure features. This configuration does not provide operator protection.
Standard Features
  • No ductwork required
  • No installation required - plugs directly into a standard 110V or 220V electrical outlet
  • Encloses equipment while retaining full operator access
  • UVTect™ Microprocessor Controller
  • ISO 5 vertical laminar flow air
  • UV and fluorescent lights
  • Seamless plastic design - no joints or gaps in construction
  • Lab event timer
  • HEPA filtration for capture of particulate to 0.3 microns in size
Unique Safety Features
  • Filter safety lockout to ensure correct filter replacement
  • Polycarbonate and polypropylene design reflects UV energy
  • Tested to UL 61010-1,CSA C22.2 No. 61010-1, and EN 61326-1
  • Brushless, sparkless blower is post-filter
  • Filter and electronics are located above the workspace, eliminating the chance of chemical spill
  • Real time monitoring of installed filtration bed
Specifications
Product Type Microscope Enclosure
Includes UVTect™ Microprocessor Controller
Width (English) 32 in.
Width (Metric) 813 mm