Learn More
Hamilton AirShield
Product Code. 31257782
Change view
Click to view available options
For Use With (Equipment):
Microlab 600 Instrument
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment)
31257782 Microlab 600 Instrument
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31257782 Supplier Hamilton Supplier No. 6856201
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AirShield accessory designed for integration with compatible laboratory systems. Supports controlled operation and protection during use.
  • Airshield is an optional accessory for Microlab 600 instruments
  • Creates a positive-pressure airflow inside the pump
  • Directs air over critical components and out of the system
  • Helps protect internal components from environmental contamination
Specifications
Product Type AirShield
For Use With (Equipment) Microlab 600 Instrument