missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ajuba Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92766-0.1ml
This item is not returnable.
View return policy
Description
Ajuba Polyclonal antibody specifically detects Ajuba in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Ajuba | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| Ajuba, jub, ajuba homolog (Xenopus laevis), MGC15563, protein ajuba | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Ajuba (NP_116265.1). MERLGEKASRLLEKFGRRKGESSRSGSDGTPGPGKGRLSGLGGPRKSGPRGATGGPGDEPLEPAREQGSLDAERNQRGSFEAPRYEGSFPAGPPPTRALP | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 84962 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction