Learn More
AKIP1 Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808486S
Description
AKIP1 Monoclonal antibody specifically detects AKIP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| AKIP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15633 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6895 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI2E4 | |
| Unconjugated | |
| TARBP2 | |
| A kinase (PRKA) interacting protein 1; A-kinase interacting protein 1; A-kinase-interacting protein 1; Akip1; BCA3; breast cancer associated 3; breast cancer associated gene 3; breast cancer-associated gene 3 protein; C11orf17; D7H11orf17; D930014E17Rik; ICRFP703B1614Q5.6; ICRFP703N2430Q5.6; koyt binding protein 1; koyt binding protein 2; koyt binding protein 3; ORF27; PKA-interacting protein; Proline-rich protein BCA3; protein kinase A-interacting protein 1; RGD1306959 | |
| Full length human recombinant protein of human TARBP2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.