missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523602
This item is not returnable.
View return policy
Description
AKIP1 Polyclonal Antibody for Western Blot
Enhances NF-kappa-B transcriptional activity by regulating the nuclear localization of the NF-kappa-B subunit RELA and promoting the phosphorylation of RELA by PRKACA. Regulates the effect of the cAMP-dependent protein kinase signaling pathway on the NF-kappa-B activation cascade.
Specifications
| AKIP1 | |
| Polyclonal | |
| Unconjugated | |
| Akip1 | |
| A kinase (PRKA) interacting protein 1; A-kinase interacting protein 1; A-kinase-interacting protein 1; Akip1; BCA3; breast cancer associated 3; breast cancer associated gene 3; breast cancer-associated gene 3 protein; C11orf17; D7H11orf17; D930014E17Rik; ICRFP703B1614Q5.6; ICRFP703N2430Q5.6; koyt binding protein 1; koyt binding protein 2; koyt binding protein 3; ORF27; PKA-interacting protein; Proline-rich protein BCA3; protein kinase A-interacting protein 1; RGD1306959 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100751192, 56672, 57373 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9JJR5, Q9NQ31 | |
| Akip1 | |
| KLH conjugated synthetic peptide between 54-83 amino acids from the N-terminal region of human BCA3. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Hamster | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction