missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKNA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23785-1-AP-20UL
This item is not returnable.
View return policy
Description
AKNA is a transcription factor that specifically activates the expression of the CD40 receptor and its ligand CD40L/CD154, two cell surface molecules on lymphocytes that are critical for antigen-dependent-B-cell development. It binds to A/T-rich promoters.Specifications
| AKNA | |
| Polyclonal | |
| Unconjugated | |
| AKNA | |
| AKNA, AT hook transcription factor, KIAA1968, RP11 82I1.4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80709 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z591 | |
| AKNA | |
| AKNA Fusion Protein Ag20662 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction