missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Multispecies AKT/ERK Activation InstantOne™ ELISA Kit

Product Code. 15580767
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15580767 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15580767 Supplier Invitrogen™ Supplier No. 858601411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster ERK 1/2 and AKT 1/2/3 in cell lysates. Detection of rat ERK 1/2 and AKT 1/2/3 is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

Extracellular signal-regulated kinase (ERK) is the founding member and a key component of the classical Mitogen-Activated Protein Kinase (MAPK) pathway. ERK1 (p44, MAPK3) and ERK2 (p42, MAPK2) are both activated via the MAPK/ERK pathway, which is downstream of various Receptor Tyrosine Kinases (RTKs) or GPCRs. MEK, a Ser/Thr and Tyr kinase, activates ERK by phosphorylation of its TxY motifs, namely Thr202/Tyr204 and Thr185/Tyr187 of ERK1 and ERK2, respectively. Once activated, ERK phosphorylates a variety of proteins that regulate cellular processes such as cell division, proliferation, survival, differentiation, apoptosis, motility, and metabolism. AKT, which exists as multiple isoforms, is one of the principle kinases activated by phosphoinositide 3-kinase (PI3K). Activation of PI3K results in the generation of phosphatidylinositol (3,4,5)-triphosphate (PIP3). These lipid second messengers bind to the pleckstrin homology domain of AKT to promote its translocation to the plasma membrane for activation via phosphorylation at Thr308 and Ser473 by PDK1 and the mTOR TORC2 complex, respectively. Phosphorylation at both these sites is required for full activation of AKT Ser/Thr kinase activity. AKT phosphorylates over 50 known substrates, including GSK3, AS160, PRAS40, TSC 1, TCS 2, Raf-1, Bad, the FOXO family of transcription factors, and PFK2. Due to its established critical role in numerous biological processes, AKT is a frequent target for oncology-related drug development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27361,P28482,P31749,P31751,Q9Y243
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 10000, 207, 208, 5594, 5595
Gene Symbol AKT,CWS6,PKB,PKB-ALPHA,PRKBA,RAC,RAC-ALPHA,ERK,ERK-2,ERK2,ERT1,MAPK2,P42MAPK,PRKM1,PRKM2,p38,p40,p41,p41mapk,p42-MAPK,ERK-1,ERK1,ERT2,HS44KDAP,HUMKER1A,P44ERK1,P44MAPK,PRKM3,p44-ERK1,p44-MAPK
Kit Contents Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 20 μL
Storage Requirements 2°C to 8°C
Synonym PKB, RAC-Pk.-alpha / p44/p42, MAPK3/MAPK2
Target AKT Pan/Erk1/2
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 10 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.