missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1/2/3 Antibody (SR2006), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00€ - 449.00€
Specifications
| Antigen | AKT1/2/3 |
|---|---|
| Clone | SR2006 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642445
|
Novus Biologicals
NBP3-21678-100ul |
100 μg |
449.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651735
|
Novus Biologicals
NBP3-21678-50ul |
50 μg |
349.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKT1/2/3 Monoclonal antibody specifically detects AKT1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| AKT1/2/3 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AKT, EC 2.7.11, EC 2.7.11.1, PKBMGC99656, PRKBA, Protein kinase B, Proto-oncogene c-Akt, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, v-akt murine thymoma viral oncogene homolog 1 | |
| A synthesized peptide derived from human AKT1/2/3 (Uniprot #: P31749, P31751, Q9Y243) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2006 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 207 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title