missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1 Antibody (CL0887) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44500
This item is not returnable.
View return policy
Description
AKT1 Monoclonal antibody specifically detects AKT1 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| AKT1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, Serine-Threonine Protein Kinase, v-akt murine thymoma viral oncogene homolog 1, V-Akt Murine Thymoma Viral Oncogene-Like Protein 1 | |
| This antibody was generated using a recombinant protein sequence of P31749, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Angiogenesis, Apoptosis, Autophagy, Cancer, Cellular Markers, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Serine Kinases, Signal Transduction, Tyrosine Kinases | |
| 207 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL0887 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction