missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aflatoxin B1 Antibody (6A10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 21 publication
226.00€ - 530.00€
Specifications
| Antigen | Aflatoxin B1 |
|---|---|
| Clone | 6A10 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:500-1:2000, ELISA 5 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 24114584), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24391771), Immunohistochemistry-Frozen, Dot Blot reported in scientific literature (PMID 19524575) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18283925
|
Novus Biologicals
NB600-443SS |
0.025 mL |
226.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18757433
|
Novus Biologicals
NB600-443 |
0.1 mL |
530.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aflatoxin B1 Monoclonal specifically detects Aflatoxin B1 in Human, All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.Specifications
| Aflatoxin B1 | |
| 1.0 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| AFB1, AFB1-AR1, Aldoketoreductase 7 | |
| IgG1 κ | |
| Protein G purified | |
| All species. There is no cross-reactivity with other carcinogen-DNA adducts. |
| 6A10 | |
| Western Blot 1:500-1:2000, ELISA 5 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 24114584), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24391771), Immunohistochemistry-Frozen, Dot Blot reported in scientific literature (PMID 19524575) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, All species | |
| Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title