Learn More
Al-23 Protective Tube, One End Closed, O.D.: 20mm, I.D.: 15mm, Wall.: 2.5mm, Thermo Scientific™
Product Code. 11382698
Change view
Click to view available options
Quantity:
200 mm
270 mm
375 mm
530 mm
740 mm
1030 mm
1430 mm
2030 mm
Unit Size:
1030mm
1430mm
200mm
2030mm
270mm
375mm
530mm
740mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382698 1030 mm 1030mm
11362698 200 mm 200mm
11332698 270 mm 270mm
11352698 375 mm 375mm
11392698 530 mm 530mm
11342698 740 mm 740mm
11302708 1430 mm 1430mm
11372698 2030 mm 2030mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11382698 Supplier Thermo Scientific Alfa Aesar Supplier No. 030052.EQ
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Protective Tube
Name Note One end closed
Quantity 1030 mm