missing translation for 'onlineSavingsMsg'
Learn More

ALDH1L2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

190.00€ - 463.00€

Specifications

Antigen ALDH1L2
Dilution Western Blot 1:1000 - 1:5000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18641361
View Documents
Novus Biologicals
NBP2-92166-0.02ml
0.02 mL
190.00€
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18604651
View Documents
Novus Biologicals
NBP2-92166-0.1ml
0.1 mL
463.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ALDH1L2 Polyclonal antibody specifically detects ALDH1L2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

ALDH1L2
Western Blot
Unconjugated
Rabbit
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
PBS with 50% glycerol, pH7.3.
160428
IgG
Affinity purified
Western Blot 1:1000 - 1:5000
Polyclonal
Purified
RUO
Human, Mouse, Rat
aldehyde dehydrogenase 1 family, member L2, aldehyde dehydrogenase family 1 member L2, DKFZp686A16126, DKFZp686M064, EC 1.5.1.6, FLJ36769, FLJ38508, MGC119536, MGC119537,10-formyltetrahydrofolate dehydrogenase ALDH1L2, Mitochondrial 10-formyltetrahydrofolate dehydrogenase, mtFDHDKFZp686P14145, probable 10-formyltetrahydrofolate dehydrogenase ALDH1L2
Recombinant fusion protein containing a sequence corresponding to amino acids 280-480 of human ALDH1L2 (NP_001029345.2). EPLEIKGAKKPGLVTKNGLVLFGNDGKALTVRNLQFEDGKMIPASQYFSTGETSVVELTAEEVKVAETIKVIWAGILSNVPIIEDSTDFFKSGASSMDVARLVEEIRQKCGGLQLQNEDVYMATKFEGFIQKVVRKLRGEDQEVELVVDYISKEVNEIMVKMPYQCFINGQFTDADDGKTYDTINPTDGSTICKVSYASLA
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.