missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH2 Antibody (4G6A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
217.00€ - 552.00€
Specifications
| Antigen | ALDH2 |
|---|---|
| Clone | 4G6A3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18678195
|
Novus Biologicals
NBP2-37397-0.025ml |
0.025 mL |
217.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18166088
|
Novus Biologicals
NBP2-37397 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALDH2 Monoclonal specifically detects ALDH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| ALDH2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| acetaldehyde dehydrogenase 2, aldehyde dehydrogenase 2 family (mitochondrial), aldehyde dehydrogenase, mitochondrial, ALDH class 2, ALDH-E2, ALDHI, ALDM, EC 1.2.1, EC 1.2.1.3, liver mitochondrial ALDH, MGC1806, nucleus-encoded mitochondrial aldehyde dehydrogenase 2 | |
| ALDH2 | |
| IgG1 | |
| Affinity Purified |
| 4G6A3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat | |
| 217 | |
| Purified recombinant fragment of human ALDH2 (AA: 317-517) expressed in E. Coli. | |
| Primary | |
| 56.3 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title