missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH7A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10368-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of subfamily 7 in the aldehyde dehydrogenase gene family. These enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular member has homology to a previously described protein from the green garden pea, the 26g pea turgor protein. It is also involved in lysine catabolism that is known to occur in the mitochondrial matrix. Recent reports show that this protein is found both in the cytosol and the mitochondria, and the two forms likely arise from the use of alternative translation initiation sites. An additional variant encoding a different isoform has also been found for this gene. Mutations in this gene are associated with pyridoxine-dependent epilepsy. Several related pseudogenes have also been identified.Specifications
| ALDH7A1 | |
| Polyclonal | |
| Unconjugated | |
| ALDH7A1 | |
| ALDH7A1, Alpha AASA dehydrogenase, Antiquitin 1, ATQ1, Betaine aldehyde dehydrogenase, EPD, P6c dehydrogenase, PDE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110695, 291450, 501 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49419, Q64057, Q9DBF1 | |
| Aldh7a1 | |
| ALDH7A1 Fusion Protein Ag0383 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction