missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Aldolase A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30648
This item is not returnable.
View return policy
Description
Aldolase A Polyclonal antibody specifically detects Aldolase A in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Aldolase A | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ALDA, aldolase A, fructose-bisphosphate, EC 4.1.2.13, fructose-1,6-bisphosphate triosephosphate-lyase, fructose-bisphosphate aldolase A, GSD12, Lung cancer antigen NY-LU-1, MGC10942, MGC17716, MGC17767, Muscle-type aldolase | |
| Between 325 and 375 | |
| 100 μg | |
| Cancer, Cell Cycle and Replication | |
| 226 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction