Learn More
Invitrogen™ Aletekutug Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558332
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Interleukin-18 (IL-18) is a cytokine belonging to the IL-1 family and is initially synthesized as an inactive precursor, pro-IL-18. This precursor undergoes proteolytic processing by caspase-1, leading to the production of mature IL-18. Structurally, mature IL-18 forms a complex with its receptor IL-18R alpha and, along with the accessory protein IL-18R beta, initiates a signaling cascade. Functionally, IL-18 plays a crucial role in both innate and adaptive immunity by inducing the production of interferon-gamma (IFN-gamma), particularly in concert with IL-12, which activates natural killer (NK) cells and promotes Th1 cell responses. Additionally, IL-18 can influence Th2 and IL-17-producing gamma delta T cells, macrophage activation, and inflammatory responses, underscoring its multifaceted role in immune regulation and disease pathogenesis.
Specifications
| Aletekutug Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.