missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Alix Polyclonal antibody specifically detects Alix in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | Alix |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:5000, Simple Western, Flow Cytometry 1-5 ug/mL, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1-5 ug/mL, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin |
| Formulation | PBS with 0.09% Sodium Azide |
| Gene Alias | AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein |
| Gene Symbols | PDCD6IP |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?