missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Concentration | 15 mg/mL |
| For Use With (Application) | ELISA |
| Formulation | 70% Ammonium sulfate |
| Gene ID (Entrez) | 249 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 110 kDa |
| Quantity | 5 mg |
| Storage Requirements | Store at -20°C. |
| Gene Alias | Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935 |
| Gene Symbol | ALPL |
| Research Category | Stem Cell Markers |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?