missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH5 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-78817AF594
This item is not returnable.
View return policy
Description
ALKBH5 Polyclonal antibody specifically detects ALKBH5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ALKBH5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| ABH5OFOXD, alkB, alkylation repair homolog 5 (E. coli), Alkylated DNA repair protein alkB homolog 5, EC 1.14.11.-, FLJ20308, OFOXD1, oxoglutarate and iron-dependent oxygenase domain containing, probable alpha-ketoglutarate-dependent dioxygenase ABH5 | |
| Partial recombinant protein made to an N-terminal portion of the human ALKB5H protein (amino acids 1-394) [UniProt 54890]. | |
| 0.1 mL | |
| Direct Reversal of DNA Damage | |
| 54890 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction