Learn More
Alkyl-DHAP synthase/AGPS Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21011-1-AP-150UL
Description
The Alkyl-DHAP synthase/AGPS antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Alkyl-DHAP synthase/AGPS. This antibody recognizes human antigen. The Alkyl-DHAP synthase/AGPS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Alkyl-DHAP synthase/AGPS | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 8540 | |
| IgG |
| Western Blot, ELISA | |
| 560 μg/mL | |
| Rabbit | |
| Primary | |
| Human |