missing translation for 'onlineSavingsMsg'
Learn More

Allyl Glycidyl Ether, TRC

Product Code. 30326758
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326758 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326758 Supplier Toronto Research Chemicals Supplier No. TRCA5557001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(2-Propen-1-yloxy)methyl]oxirane
CAS 106-92-3
Molecular Formula C6 H10 O2
Quantity 1 g
Synonym Oxirane, 2-[(2-propen-1-yloxy)methyl]-, Oxirane, [(2-propenyloxy)methyl]- (9CI), Propane, 1-(allyloxy)-2,3-epoxy- (6CI,8CI), 2-[(2-Propen-1-yloxy)methyl]oxirane, (±)-Allyl glycidyl ether, 1,2-Epoxy-3-allyloxypropane, 1-(2-Propenyloxy)-2,3-epoxypropane, 1-Allyloxy-2,3-epoxypropane, 2-(Allyloxymethyl)oxirane, 2-[(2-Propenyloxy)methyl]oxirane, 2-[(Prop-2-en-1-yloxy)methyl]oxirane, A 32608, AGE, AGE 500, Allyl 2,3-epoxypropyl ether, Allyl glycidyl ether, Allyl oxiranemethyl ether, Denacol EX 111, Glycidyl 2-propenyl ether, Glycidyl allyl ether, NSC 18596, NSC 631, Silong 191, [(2-Propenyloxy)methyl]oxirane, [(Allyloxy)methyl]oxirane
InChI Formula InChI=1S/C6H10O2/c1-2-3-7-4-6-5-8-6/h2,6H,1,3-5H2
SMILES C=CCOCC1CO1
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
Molecular Weight (g/mol) 114.14
Formula Weight 114.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.