missing translation for 'onlineSavingsMsg'
Learn More

Allyltributyltin, TRC

Product Code. 30325373
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325373 5 g 5g
30325250 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30325373 Supplier Toronto Research Chemicals Supplier No. TRCA5723305G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allyltributyltin
CAS 24850-33-7
Molecular Formula C15H32Sn
Quantity 5 g
Synonym Tributyl-2-propen-1-ylstannane, (Propen-2-yl)tributyltin, 3-(Tributyltin)-1-propene, 3-Tributylstannyl-1-propene, Allyltri-n-butylstannane, Tri(n-butyl)allylstannane, Tributyl(2-propenyl)stannane, Tributylallylstannane
InChI Formula InChI=1S/3C4H9.C3H5.Sn/c3*1-3-4-2;1-3-2;/h3*1,3-4H2,2H3;3H,1-2H2;
SMILES CCCC[Sn](CCCC)(CCCC)CC=C
IUPAC Name tributyl(prop-2-enyl)stannane
Molecular Weight (g/mol) 331.12
Formula Weight 332.1526
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.