missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALOX15 Monoclonal Antibody (6#), Invitrogen™

Mouse Monoclonal Antibody
185.00€ - 465.00€
Specifications
| Antigen | ALOX15 |
|---|---|
| Clone | 6# |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17664436
|
Invitrogen™
MA543982 |
20 μL |
185.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17654436
![]() |
Invitrogen™
MA543981 |
100 μL |
465.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALOX15 converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid. The protein also acts on C-12 of arachidonate as well as on linoleic acid.Specifications
| ALOX15 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 12/15-lipoxygenase; 12/15-LO; 12/15-LOX; 12-LO; 12-LOX; 15-lipooxygenase-1; 15-LOX; 15-LOX-1; 15LOX-1; 15-LOX2; Alox12; Alox12l; Alox15; Arachidonate 12-lipoxygenase, leukocyte-type; arachidonate 15-lipoxygenase; Arachidonate omega-6 lipoxygenase; L-12LO; LOG15 | |
| Alox15 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| 6# | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P16050 | |
| 246 | |
| Lys170-Leu312 (Accession P16050), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Alox15 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title