missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Trifluoro-p-tolyl isocyanate, 99%, ACROS Organics™

Product Code. 10680054 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10680054 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10680054 Supplier Thermo Scientific Acros Supplier No. 314220010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1548-13-6
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H4F3NO
Linear Formula CF3C6H4NCO
Refractive Index 1.4680 to 1.4710 (20°C, 589nm)
MDL Number MFCD00002028
Quantity 1 g
Synonym 4-trifluoromethyl phenyl isocyanate, 1-isocyanato-4-trifluoromethyl benzene, alpha,alpha,alpha-trifluoro-p-tolyl isocyanate, 4-trifluoromethyl phenylisocyanate, 4-isocyanatobenzotrifluoride, 4-trifluoromethylphenyl isocyanate, benzene, 1-isocyanato-4-trifluoromethyl, p-trifluoromethylphenyl isocyanate, isocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethyl benzenisocyanate
InChI Key QZTWVDCKDWZCLV-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC=C(C=C1)N=C=O
IUPAC Name 1-isocyanato-4-(trifluoromethyl)benzene
Molecular Weight (g/mol) 187.12
PubChem CID 73768
Formula Weight 187.12
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluoro-p-tolyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.