missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Hydroxy Olopatadine Hydrochloride, TRC

Product Code. 17979939
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17979939 1 mg 1mg
18061220 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17979939 Supplier Toronto Research Chemicals Supplier No. TRCH9487701MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1331668-21-3
Molecular Formula C21 H23 N O4 . Cl H
Molecular Weight (g/mol) 389.87
Synonym 11-[3-(Dimethylamino)propylidene]-6,11-dihydro-α-hydroxydibenz[b,e]oxepin-2-acetic Acid Hydrochloride (1:1)
IUPAC Name 2-[11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]-2-hydroxyacetic acid;hydrochloride
SMILES Cl.CN(C)CCC=C1c2ccccc2COc3ccc(cc13)C(O)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H23NO4.ClH/c1-22(2)11-5-8-17-16-7-4-3-6-15(16)13-26-19-10-9-14(12-18(17)19)20(23)21(24)25;/h3-4,6-10,12,20,23H,5,11,13H2,1-2H3,(H,24,25);1H
Formula Weight 389.1394
Chemical Name or Material alpha-hydroxy Olopatadine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.