missing translation for 'onlineSavingsMsg'
Learn More

alpha-Ketoglutaric acid disodium salt dihydrate, 99%

Product Code. 15400297
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
250 g
Unit Size:
100g
1g
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400297 100 g 100g
15445419 1 g 1g
15490287 25 g 25g
15410297 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15400297 Supplier Thermo Scientific Alfa Aesar Supplier No. J63154.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 305-72-6 | C5H4Na2O5 | 190.062 g/mol

alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

Solubility
Soluble in water (1 M).

Notes
Hygroscopic. Store under inert gas. Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-72-6
Molecular Formula C5H4Na2O5
Molecular Weight (g/mol) 190.062
MDL Number MFCD00150702
InChI Key YBGBJYVHJTVUSL-UHFFFAOYSA-L
Synonym disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
PubChem CID 31040
IUPAC Name disodium;2-oxopentanedioate
SMILES C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (1 M).
Formula Weight 226.09 (190.06 Anhydrous)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material alpha-Ketoglutaric acid disodium salt dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.