missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Phenyl-Alpha-(2-propyn-1-yloxy)-benzeneacetic Acid Methyl Ester, TRC

Product Code. 30409566
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409566 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409566 Supplier Toronto Research Chemicals Supplier No. TRCP336413250MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Phenyl-alpha-(2-propyn-1-yloxy)-benzeneacetic Acid Methyl Ester
CAS 30118-97-9
Molecular Formula C18H16O3
Quantity 250 mg
Synonym Diphenyl(2-propynyloxy)-acetic Acid Methyl Ester
InChI Formula InChI=1S/C18H16O3/c1-3-14-21-18(17(19)20-2,15-10-6-4-7-11-15)16-12-8-5-9-13-16/h1,4-13H,14H2,2H3
SMILES COC(=O)C(OCC#C)(c1ccccc1)c2ccccc2
IUPAC Name Methyl 2,2-d=Diphenyl-2-(prop-2-yn-1-yloxy)acetate
Molecular Weight (g/mol) 280.32
Formula Weight 280.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.