missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Phenyl-Alpha-(2-pyridyl)acetonitrile, TRC

Product Code. 30419948
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419948 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419948 Supplier Toronto Research Chemicals Supplier No. TRCP33665050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5005-36-7
Molecular Formula C13 H10 N2
Molecular Weight (g/mol) 194.23
Synonym 2-Pyridineacetonitrile, α-phenyl-, 2-Phenyl-2-(2-pyridyl)acetonitrile, 2-Pyridylphenylacetonitrile, NSC 16276, Phenyl(pyridin-2-yl)acetonitrile, α-Phenyl-2-pyridineacetonitrile, α-Phenyl-α-(2-pyridyl)acetonitrile
IUPAC Name 2-phenyl-2-pyridin-2-ylacetonitrile
SMILES N#CC(c1ccccc1)c2ccccn2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H10N2/c14-10-12(11-6-2-1-3-7-11)13-8-4-5-9-15-13/h1-9,12H
Formula Weight 194.0844
Chemical Name or Material a-Phenyl-a-(2-pyridyl)acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.