missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaB Crystallin/CRYAB, p Ser45 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-5598AF405
This item is not returnable.
View return policy
Description
AlphaB Crystallin/CRYAB Polyclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AlphaB Crystallin/CRYAB | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
| Synthetic phopshopeptide corresponding to residues L(44) S(p) P F Y L R P P S F(54) C of human alpha-B Crystallin. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
| 1410 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction