Description
- Silicone Free
- Low Linting
Specifications
Specifications
| Type | Chemical-resistant Apron |
| Size | Small |
| Material | Non-woven Microporous Polyethylene Laminate |
| Color | White |
| Disposable | Limited-use |
| Closure Type | Tear-back |
| Sterility | Non-sterile |
| Certifications/Compliance | EN ISO 13688:2013, Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003 |
| Weight (Metric) | 63 gsm |
| Style | W20B214 |
| Show More |