Description
- The AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and taped - Model 214 chemical protective suit is made with a multi-layer barrier fabric, which offers wearers excellent protection against several chemicals, including inorganic and biological hazards
- These highly visible, bright yellow chemical protection coveralls improve worker safety in dimly lit environments
- Featuring welded and taped seams, this full chemical protection suit offers a strong barrier against harmful liquid and particles
Specifications
Specifications
| Type | Apron |
| Size | Large |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Closure Type | Hook and Loop (Rear) |
| Sterility | Non-sterile |
| Latex Free | Yes |
| No. of Pockets | None |
| Seam Type | Welded and Taped |
| Show More |