missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 3000 Ultrasonically Welded and Taped - Model 121-G02

Product Code. 31114768 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
31114768 Small
31114769 Medium
31114770 Large
31114771 X-Large
31114772 2X-Large
31114773 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31114768 Supplier Ansell™ Supplier No. YE30T0012102G02
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical protective suits with dual-zip systems, double cuffs and robust barrier protection

  • The AlphaTec™ 3000 Ultrasonically Welded & Taped - Model 121-G02 protective suit features a multi-layered protective barrier, capable of withstanding a range of inorganic chemicals and biohazards
  • It is also designed with dual-zip systems and double cuffs, for even more robust barrier protection
  • This protective outer garment also remains highly visible in low-lit environments, for enhanced wearer and workplace safety
  • This AlphaTec™ protective suit also complies with EN 1149-5 antistatic standards
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper
Seam Type Welded, Taped
Sterility Non-sterile
Resistance Chemicals, Particle, Anti-static, Bloodborne Pathogens
Certifications/Compliance (Europe) CE CATEGORY III (Type 3-B, Type 4-B, Type 5-B, Type 6-B, EN 1073-2, EN 14126, EN 1149-5)
Latex Free Yes
No. of Pockets None
For Use With (Application) Transferring Liquids and Solids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.