Learn More
Ansell AlphaTec™ 37-210 Reusable Nitrile Gloves with UltraGrip™ Technology
Product Code. 16895200 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
12
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16895200 10 12 pairs
16865200 7 12 pairs
16875200 8 12 pairs
16885200 9 12 pairs
16805210 11 12 pairs
16815210 12 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16895200 Supplier Ansell Supplier No. 37210100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
UltraGrip™ Technology delivers an innovative raised diamond grip on the glove's surface for enhanced grip capabilities in wet and oily conditions.
  • Reusable nitrile gloves designed specifically for food processing and handling
  • Raised diamond-pattern enhances grip in wet and oily conditions
  • Gloves maintain flexibility and dexterity, even at low temperatures
  • Use alone or as outer glove over thermal liners or cut-resistant gloves
  • Latex-free formulation presents no risk of Type 1 allergies
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Color Blue
Disposable Reusable
Hand Specific
Latex Free Yes
Material Nitrile
Size 10
Thickness (English) 8 mil
EN388 Standard 3001X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKT
Series 37-210