missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 38-560 Chemical Resistant Butyl Gloves - Gauntlet Cuff

Product Code. 16855220 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16855220 8 10 pairs
16845220 7 10 pairs
16865220 9 10 pairs
16875220 10 10 pairs
16885220 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16855220 Supplier Ansell Supplier No. 38560080

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Gloves are constructed of a thick butyl material to provide a high level of chemical resistance against agressive chemicals, without sacrificing comfort or dexterity.

  • High protection level against gases, particularly ozone
  • Extended gauntlet style cuff for additional protection
  • Beaded cuff for tear resistance and easy donning
  • Rough finish on the hand for safer handling
  • Designed for comfort with a natural, curved ergonomic shape
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Applications Industrial
Color Black
Latex Free Yes
Material Butyl Polymer
Product Type Butyl Gloves
Size 8
EN388 Standard 1020X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABIKPT
EN ISO 374 - 5 Yes
Glove Cuff Style Gauntlet
Resistant To Solvents, and Strong Acids
Series 38-560
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.