Learn More
Ansell AlphaTec™ 4000 APOLLO
Descripción
- Designed to protect, rear entry double zip system, attached socks with boot overflap
- Rear mounted breathing apparatus (BA) pouch - universal fit with most BA
- Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
- Attached Ansell Barrier™ gloves, exhalation valves and clear face visor
Especificaciones
Especificaciones
| Color | Green |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
| Hood Type | Fully Encapsulated |
| Material | Multi-layer Barrier Laminate Fabric |
| Size | Small |
| Product Type | Fully encapsulated chemical protective, liquid tight suit, Type 3/4 protection |
| Seam Type | Welded and Taped |
| Style | G40T126B |
| Weight (Metric) | 103 gsm |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?