Learn More
Ansell AlphaTec™ 4000 CFR Stitched and Taped - Model 111
Product Code. 31781273 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 12
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31781273 Small Pack of 5
31781274 Medium Pack of 5
31781275 Large Pack of 5
31781276 X-Large Pack of 5
31781277 2X-Large Pack of 5
31781278 3X-Large Pack of 5
31781279 4X-Large Pack of 12
31781280 5X-Large Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 31781273 Supplier Ansell Supplier No. OR47T0011102
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Coverall with 2 piece hood, elasticated hood, waist, wrists, ankles, double zip flap closure.
  • Specialized protection: AlphaTec™ 4000 CFR Stitched and Taped - Model 111 chemical protective suit features multi-hazard solutions that offer a proven 8 hour barrier against chemicals such as non-flammable, flammable and aggressive solvents
  • Improved durability: Complying with EN ISO 14116 and ASTM D6413 Flame Resistance*, this full protective suit shields the wearer from chemical sprays without compromising safety by limiting flame spread in the event of a flash fire
  • Elevated comfort: The optimized body fit design of this protective suit offers improved wearer comfort and safety
  • This full-chemical protection suit defends against infective agents, tested according to ASTM F1671 and hydrocarbons such as hexane, HF, oleum and many more
  • *AlphaTec™ 4000 CFR should never be worn in isolation for flame retardant protection.
  • Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above
  • Polyolefin-based core barrier laminate, with FR non-woven inner and FR outer barrier film
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical-resistant Coveralls
Size Small
Gender Unisex
Material Polyolefin
Color Orange
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Hood Type Attached
Closure Type Double Zipper Flap
Seam Type Stitched, Taped
Sterility Non-sterile
Resistance Chemicals, Flame, Infectious Agents
Collar Type 2-piece Hood
Back Style Elastic Waist at Back
Certifications/Compliance EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, CE 0598, EN ISO 14116:2015 Index 1, EN ISO 13688:2013, EN 1073-2:2002, EN 1149-5:2018, EN 14126:2003
Latex Free Yes
No. of Pockets None
For Use With (Application) Fire and Rescue, Haz-mat response, Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Raw material sample collection, Concentration mixing and palletisation, Assembly and disassembly of large pipes