missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ CFR
Description
- Multi hazard protection from chemical spray without compromising wearer safety in the event of a flash fire
- Flame retardant treated fabric with PVC barrier film protection
- Offers protection from pressurised liquid spray to EN Type 3 and 4 and particulates to EN Type 5
- Optimized body fit, improving wearer comfort and safety
Specifications
Specifications
| Size | 3X-Large |
| Material | FR Treated Sontara, Wood Pulp and Polyester with PVC Barrier Film |
| Color | Red |
| Seam Type | Stitched, Taped |
| Certifications/Compliance (Europe) | EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2018, EN 14126:2003, EN ISO 13688:2013, EN ISO 14116:2015 |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
| Weight (Metric) | 165 gsm |
| Style | R96T103 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?