missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR3 Antibody (S390-43), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59333-25ug
This item is not returnable.
View return policy
Description
ALS2CR3 Monoclonal specifically detects ALS2CR3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ALS2CR3 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.1% Sodium Azide | |
| TRAK2 | |
| Fusion protein amino acids 757-914 (C-terminus) of human TRAK2 | |
| 25 μg | |
| Primary | |
| Detects 100kDa. Does not cross-react with TRAK1. | |
| Store at -20°C. | |
| IgG2a |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| S390-43 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| ALS2CR3GRIF1, CALS-C, GRIF-1, KIAA0549amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3, OIP98Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, trafficking kinesin-binding protein 2, trafficking protein, kinesin binding 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 66008 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction