Learn More
Euro Protection™ Altaite Low-Cut Safety Shoes
Product Code. 12083676
Change view
Click to view available options
Size:
37
40
41
42
43
44
45
47
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12083676 43 Each
12023676 37 Each
12053676 40 Each
12063676 41 Each
12073676 42 Each
12093676 44 Each
12003686 45 Each
12023686 47 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12083676 Supplier Euro Protection™ Supplier No. 9ALTL43
  • ONLINE EXCLUSIVE PRICE
    94.55€
    48.65€ / Pack of 1
    Save 45.90€
    48% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Euro Protection™ Altaite Low-Cut Safety Shoes are comfortable, snug, and durable, and feature a composite toe cap for protection.
  • Padded collar and bellows tongue for snug fit.
  • Heel scuff cap
  • Shock-absorbing EVA midsole.
  • Full grain leather and bright mesh combination upper.
  • Breathable 3D mesh lining.
  • Removable, textile and foam insole.
Specifications
Color Brown, Beige
Liner Material Breathable, 3D Mesh
Material Leather
Size 43
Sole Features Cemented, shock-absorbing EVA midsole, nitrile rubber outsole, non-metallic (composite fabric) puncture-resistant sole.
Sole Material Nitrile, EVA, Composite Fabric
Style Low
Toe Material Composite
Weight (Metric) 603 g per shoe in size 8
Certifications/Compliance (Europe) EN ISO 20345 : 2011