Learn More
Macherey-Nagel™ Alugram Aluminum Sheet CEL 300
Product Code. 10368601 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
4 x 8 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10368601 20 x 20 cm Pack of 25
10328651 4 x 8 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10368601 Supplier Macherey-Nagel™ Supplier No. 818153
  • ONLINE EXCLUSIVE PRICE
    171.00€
    165.00€ / Pack of 25
    Save 6.00€
    3% Off
View all promotions
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Quantity 25 Pk.
Type Aluminum Sheet
For Use With (Application) Partition Chromatography Of Polar Substances Such As Amino Acids, Carboxylic Acids Or Carbohydrates
Specific Surface 15,000 cm2/g
Dimensions (L x W) 20 x 20 cm