missing translation for 'onlineSavingsMsg'
Learn More

Alumina N, 32 to 63 μm, Non-active, MP EcoChrome™

Product Code. 17887024 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17887024 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17887024 Supplier MP Biomedicals Supplier No. 0202061
  • ONLINE EXCLUSIVE PRICE
    74.95€
    71.40€ / 500g
    Save 3.55€
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

An amphoteric oxide, commonly referred to as alumina (α-alumina), aloxide, or corundum in its crystalline form

MP EcoChrom™ Alumina N is an oxide of aluminum, occurring in nature as various minerals such as bauxite, corundum, etc.

It is used as:

  • An adsorbent
  • A desiccating agent
  • A catalyst
  • In the manufacture of dental cements and refractories

Application Notes:

MP EcoChrom™ Alumina N catalyses a variety of reactions that are useful industrially.

It is widely used:

  • To remove water from gas streams
  • For its hardness and strength
  • As an abrasive, including as a much less expensive substitute for industrial diamond
    • Many types of sandpaper use aluminium oxide crystals
  • In chemistry laboratories, alumina is a medium for chromatography, available in basic (pH 9.5), acidic (pH 4.5 when in water) and neutral formulations, as well as for:
    • Alcohols
    • Hydrocarbons
    • Steroids
    • Alkaloids
    • Natural pigments
    • Neutral alumina (less active than the basic form) is useful for separation of:
      • Aldehydes
      • Ketones
      • Quinones
      • Esters
      • Lactones
      • Glucosides
    • Acidic alumina, is the weakest and least often used type and is useful for separation of acid pigments and strong acids. It can cause:
      • Polymerization
      • Condensation
      • Dehydration reactions
    • Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification.
  • A dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties

Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Chromatography
  • Adsorbants
  • TLC

Specifications

Boiling Point ca. 2980°C (Lit.)
Melting Point ca. 2072°C
CAS 1344-28-1
Color White
pH 7.4
Physical Form Powder
Quantity 500 g
Molecular Formula Al2O3
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminium(3+) trioxidandiide
Molecular Weight (g/mol) 101.96
Density 0.8 g/mL
Chemical Name or Material Aluminum oxide
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.