Learn More
Heinz Herenz Aluminium Applicator Single Cotton Bud
Product Code. 10200713
Change view
Click to view available options
Tip Type:
Single End
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Type Unit Size
10200713 Single End Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10200713 Supplier Heinz Herenz Supplier No. 1032648
  • ONLINE EXCLUSIVE PRICE
    26.06€
    19.49€ / Pack of 100
    Save 6.57€
    25% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Aluminum applicator with single cotton bud.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
Specifications
Autoclavable Not Autoclavable
Diameter (Metric) Inlet/Outlet Tube 10.5 to 12.5mm
Sterility Non-sterile
Material Glass
Product Type Applicator Stick
Tip Material Cotton
Tip Type Single End