Learn More
Aluminon
Product Code. 10548090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10548090 100 g Glass bottle 100g
10090610 25 g Glass bottle 25g
10530994 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10548090 Supplier Thermo Scientific Acros Supplier No. 103061000
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 569-58-4 | C22H23N3O9 | 473.438 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 569-58-4
Molecular Formula C22H23N3O9
Molecular Weight (g/mol) 473.438
MDL Number MFCD00040925
InChI Key AIPNSHNRCQOTRI-UHFFFAOYSA-N
Synonym Aurintricarboxylic acid, triammonium salt
PubChem CID 54729869
ChEBI CHEBI:87398
IUPAC Name triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
SMILES C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]
Specifications
Melting Point 220.0°C to 225.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Suitability suitable for determination of aluminum
Beilstein 10, IV, 4161
Merck Index 15, 318
Color Red
Solubility Information Solubility in water: 800g/L (20°C). Other solubilities: slightly soluble in ethanol, insoluble in petroleum ether
Quantity 100 g
Physical Form Crystalline Powder
Formula Weight 473.44
Chemical Name or Material Aluminon
Show More Show Less
What is the solubility of Aluminon?
Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.